Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Liquid transfer system, Costar® Transtar-96®
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Corning® Costar®
734-4004EA 0 INR InStock
CORN7605
Liquid transfer system, Costar® Transtar-96®
The Costar® Transtar-96® system is a portable, autoclavable liquid handling system designed for use with 96 well plates.
  • Sterile 96 tip disposable cartridge enables liquids to be aspirated, transferred and dispensed over a volume range of 25 to 200 µl in five µl increments
  • Ideal for changing cell culture media and screening monoclonal antibodies
  • Accuracy is rated at ±5% at all volume levels
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms