Sodium dodecyl sulphate (SDS)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Lauryl sulphate sodium salt
, Dodecyl sulphuric acid sodium salt
, Sodium lauryl sulphate
, Dodecyl sulphate sodium salt
A7249.5000EA
0
INR
A7249.5000
A7249.1000
A7249.0500
Sodium dodecyl sulphate (SDS)
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na MW: 288.38 g/mol Melting Pt: 204…207 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,II Merck Index: 14,08636 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...