Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Magnetic stirrer, TK15
Supplier: Kartell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
KART2234EA 0 INR InStock
KART2234
Magnetic stirrer, TK15
These magnetic stirrers are constructed with tested epoxy-coated metal structure.
  • Very good speed control even at low revolution
  • Chemical and corrosion resistant
  • Used with a thermostatic water tank
  • High strength permanent crusher magnet is used
These can hold 15 containers with a maximum diameter of 64 mm. The stirrer remains cool, even after continuous process, this feature makes them suitable for microbiology and biochemistry applications.
Maximum stirring capacity of the TK15 is 15 beakers of 250 ml.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms