Tongue depressor
Supplier: Kartell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KART293EA 0 INR
KART293
Tongue depressor
Tongue depressor made of PMMA (Plexiglas).
  • Transparent
  • High mechanical resistance
  • Autoclavable at 90 °C