Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

HotMasterMix (2,5X)

Supplier: Quantabio

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

733-2474EA 91250 INR InStock
QUNT2200410
HotMasterMix (2,5X)
Nucleic Acid Reagents End-point PCR Enzymes and Kits
HotMasterMix is a ready to use 2,5X reagent mix for performing high throughput PCR (based on hot start/cold stop technology). The HotMasterMix guarantees the highest specificity and consistency. The key component of the mix is the HotMaster Taq DNA polymerase that consists of a pre-mixed HotMaster inhibitor that binds and blocks the substrate-binding site of HotMaster Taq in a temperature-dependent manner. At temperatures above 60 °C, the inhibitor is released and the HotMaster Taq becomes activated (hot start). At temperatures below 60 °C, the inhibitor inactivates the Taq polymerase (cold stop) preventing non specific primer annealing or primer-dimer formation. Unlike many conventional hot start PCR techniques, such binding-releasing equilibrium is reversible throughout the PCR cycle.

  • Highly specific PCR
  • Ready to use master mix format
  • Convenient storage at 4 °C eliminates freeze and thaw cycles
  • Minimal optimisation with self adjusting Mg²⁺ buffer technology

Product comprises: Taq DNA polymerase (1,0 U/50 µl reaction), KCl, Mg²⁺ and dNTPs.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms