Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Chloroform ≥99.8% (by GC, corrected for water content) stabilised, ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: MACRON AVANTOR BRAND

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

Management of Change (MOC) category = N

4443-10EA 0 INR InStock
4443-10 MACR4443-06
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=96870c784520df552dcde9a71610c883a1c71b50&prid=web&clid=SAPDAM
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CHCl₃) (by GC, corrected for water) ≥99.8 %
ACS - Acetone, Aldehydes (as (CH₃)₂CO) Passes Test
ACS - Acid and Chloride Passes Test
ACS - Color (APHA) ≤10
ACS - Free Chlorine Passes Test
ACS - Lead (Pb) ≤0.050 ppm
ACS - Residue after Evaporation ≤0.0003 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Suitability for Use in Dithizone Tests Passes Test
ACS - Water (H₂O) ≤0.02 %
ACS - Ultraviolet Absorbance (1.00-cm cell vs. water): Passes Test
ACS UV Absorbance - 245 nm ≤1.00
ACS UV Absorbance - 254 nm ≤0.15
ACS UV Absorbance - 255 nm ≤0.25
ACS UV Absorbance - 260 nm ≤0.15
ACS UV Absorbance - 265 nm ≤0.05
ACS UV Absorbance - 270 nm ≤0.05
ACS Absorbance 280 nm ≤0.01
ACS Absorbance 300 nm ≤0.01
ACS Absorbance - 350 nm ≤0.005
ACS Absorbance - 350 - 400 nm ≤0.01
ACS - Absorbance Passes Test
Appearance (clear, colorless liquid)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms