Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Urine collection swabs, UriSwab®
Supplier: COPAN Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
COPA800CEA 0 INR InStock
COPA800C
Urine collection swabs, UriSwab®
Swabs for the sampling of urine specimens from the collection site to the testing laboratory.
  • Use directly onto the sponge or dip in the sample
  • Chemically bonded preservatives
  • Activation on contact with urine
  • Specimen can be manually extracted by shaking the device
Advanced system specifically designed for the collection, transport and preservation of urine.
According to the testing executed as indicated in the second edition of the "Quality Control of Microbiological Transport Systems" standard, in the laboratory, UriSwab® can be used to process urine specimens for the cultivation of uropathogenic bacteria and yeasts.
Sample and device storage temperature: 2 to 25 °C
Certifications: Compliance with CLSI M40-A2 standard: Ch. 9.2 Urine Culture Containers With Preservative.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms