Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

VWR®, Bovine serum albumin (BSA)

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


0332-25GEA 6270 INR InStock
0332-25G 0332-500G
VWR®, Bovine serum albumin (BSA)
Proteins and Peptides
Heat shock isolated bovine serum albumin (BSA) suitable for use in a wide range of molecular biology applications, including Western blotting.

  • Nuclease-free
  • Protease-free
  • Heavy metals (as Pb) <0,001%
  • Iron <0,0005%
  • Purity >98%
  • Powder form

A standard grade albumin for basic applications, produced by a heat-shock process in caprylic acid.

pH (5%, water) @ 25 °C 6,5 to 7,5.


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms