Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Centrifuge Tubes, Large Volume
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
525-0505EA 10671 INR InStock
VWRI525-0505 VWRI525-0506
VWR®, Centrifuge Tubes, Large Volume
PP, conical with purple HDPE flat cap and integral seal.
  • For standard centrifuge rotors
  • Free from RNase and DNase and non pyrogenic
  • Sterile and non sterile available
  • Tubes fit standard storage racks
  • For cushion type 734-0988
  • Sustainable design based on virgin resins
Perfect for clarifying large volume solutions, or concentrating live cells in tissue culture. Resins are pre-tested, and are free of oleamide and pthalates. These tubes come in a unique balanced pack of two tubes. The tubes include attached screw caps with integral elastomeric sealing ring that eliminates leaking.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms