Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
AMRE0132
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
0132-500MLEA 4017 INR InStock
VWRC0132-500ML
AMRE0132
The acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions, and ready to use solutions for customised polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids.
  • VWR’s Acryl-40â„¢ solution is a premixed stock solution for customised polyacrylamide gel electrophoresis of proteins
  • Acryl-40â„¢ is a 40% (w/v) acylamide solution
  • Liquid stable solution eliminates handling hazardous powdered acrylamide
Ultra-pure (> 99,9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimise the handling of neurotoxic acrylamide.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms