Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
LONZ17-518
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
733-1807EA 0 INR InStock
LONZ17-518Z LONZ17-518L
LONZ17-518
Gentamicin sulphate is a broad spectrum bactericidal, with activity against gram negative and gram positive organisms including Pseudomonas. It is stable over a wide range of pH and temperature and can be autoclaved. It’s normal appearance may range from nearly colourless to amber or yellow.
Recommended concentration for use in tissue culture media:
50 mg – 1 ml per litre
10 mg – 5 ml per litre
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms