Tetracycline hydrochloride USP

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

0422-25GEA 5000 INR
0422-25G
Tetracycline hydrochloride USP
Tetracycline hydrochloride

Blocks the binding of tRNA to the 30S subunit. Recommended working concentration: 15 μg/ml

Formula: C₂₂H₂₅ClN₂O₈
MW: 480,9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8
UN: 0000

Order Now

Specification Test Results

4-Ephianhydrotetracycline 2.0 %
Crystallinity PASS
Expiration Date REPORT
Heavy Metals 0.005 %
Identification PASS
Loss on Drying 2.0 %
pH (1%, Water) @25C 1.8 - 2.8
Potency (Anhydrous) 900 mcg/mg
Potency (As Is) REPORT
Specific Rotation -255 to -240 °

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR