Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Digital anemometer/hygrometer/thermometer, 445
Supplier: Testo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
628-3203EA 0 INR InStock
TEST0563.4450
Digital anemometer/hygrometer/thermometer, 445
This service instrument is s real multi-talent: the 445 has two probe inputs for the connection of a broad range of probes for measurements on ventilation/air conditioning systems. The instrument records flow velocity values in a duct, at a duct outlet or on extraction systems. In addition to this, the allrounder measures or calculates the parameters volume flow, Indoor Air Quality, temperature, relative humidity, dewpoint, absolute humidity, degree of humidity, enthalpy, pressure and CO.
  • Point and timed mean value calculation
  • Direct display of the volume flow
  • Automatic allocation of duct cross-section area to measurement site (max. 99 measurement sites)
  • Internal measurement value store (3000 measurement values)
  • PC software for the analysis, archiving and documentation of the measurement values (option)
  • Measurement of up to 6 measurement parameters simultaneously
Delivery information: Supplied with battery and calibration protocol.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms