Hydrochloric acid 37% NF

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Muriatic acidHydrochloric acid solution

0369-100MLEA 4491 INR
0369-100ML
Hydrochloric acid 37% NF
Hydrochloric acid
Formula: HCl
MW: 36,46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Heavy Metals 0.0005 %
Purity 36.5 - 38.0 %
Residue on Ignition 0.008 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR