E-64
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J586-5MGEA 10341 INR
VWRCJ586-5MG
E-64
An irreversible inhibitor of cysteine proteases like papain, calpain, and cathepsin B, H, L and S. Also an effective inhibitor of collegenase.
  • Purity (HPLC)- ≥98%