Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Lactose monohydrate ACS

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: LactobioseMilk sugar

J815-5KGEA 15439 INR InStock
J815-5KG
Lactose monohydrate ACS
Lactose monohydrate


Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360,32 g/mol
Melting Pt: 223 °C
Storage Temperature: Ambient
CAS Number: 10039-26-6
EINECS: 200-559-2
UN: 0000
Order Now
Specification Test Results
Dextrose Pass
Heavy Metals (as Pb) ≤0.0005%
Insoluble Matter ≤0.005%
Iron ≤5 ppm
Residue after Ignition ≤0.03%
Sucrose Pass
Water (KF) 4 - 6%

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms