Constructed from clear Acrylic® polymer to give complete visibility of samples at all times. Each side panel has 2×10 mm Ø plugged ports for the introduction of either gases or cables. The incubator does not have a base so that it can be placed directly over complete instruments (base is available as an accessory). Forced air circulation and electronic temperature control ensure accurately maintained conditions.
- Full visibility of samples
- Easy access to working chamber via hinged front door panel
- Choice of models with analogue or digital control
For analogue model SI60, setting the temperature is via a calibrated knob, digital model SI60D, setting the temperature is more accurately achieved via the LCD display. The latter also gives a constant reading of actual temperature. Both models have a push switch pre-set at 37°C. Additionally there is a temperature safety cut-out set at 72°C. Ideal for plant propagation, humidity tests, simulation of tropical conditions and incubation of complete instruments.
Ordering information: Supplied flat packed, for assembly at point of use. Assembly is simple and requires only a screwdriver. Optional acrylic base plate and plastic coated shelf/rack system must be ordered separately.