Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Round bottom flasks with ground joints
Supplier: Cowie
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
COWI040.0050.29EA 0 INR InStock
COWI040.0050.29 COWI040.025.19 COWI040.0010.19 COWI040.025.24 COWI040.0100.29 COWI040.0050.24 COWI040.0025.24
Round bottom flasks with ground joints
PTFE, white.
  • Machine finished, inert and usable up to 280°C
  • Tapers match standard ground glass joints
  • Do not require sealing rings
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms