Hydroxylammonium chloride, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Hydroxylamine hydrochloride
, Hydroxylamine HCl
2196-01EA
33405.21
INR
2196-01
Hydroxylammonium chloride, BAKER ANALYZED® ACS, J.T.Baker®
Hydroxylammonium chloride
Formula:
NH₂OH·HCl MW: 69,49 g/mol Boiling Pt: 306 °C (1013 hPa) Melting Pt: 151 °C Density: 1,67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00051089 CAS Number: 5470-11-1 EINECS: 226-798-2 UN: 2923 ADR: 8,III Merck Index: 13,04853 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (NH₂OH·HCl) (by KMnO₄ titrn) | ≥96.0 % |
Clarity of Alcohol Solution | Passes Test |
Residue after Ignition | ≤0.050 % |
Titrable Free Acid (meq/g) | ≤0.25 |
Ammonium (NH₄) | Passes Test |
Sulfur Compounds (as SO₄) | ≤0.005 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Trace Impurities - Mercury (Hg) | ≤0.050 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...