Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Autoclaves, cylindrical section, Osprey
Supplier: Jencons
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
JENC115-068EA 0 INR InStock
JENC115-068 JENC115-069 JENC115-066 JENC115-067 JENC115-065 JENC115-271 JENC115-272 JENC115-071 JENC115-072 JENC115-070
Autoclaves, cylindrical section, Osprey
Osprey autoclaves have cylindrical chambers and versatile incorporate a wide variety of features to suit most requirements.
  • Protection against low water level is provided by cut-out device
  • Choice of microprocessor control system
Steam is generated by heating water in a reservoir within a chamber. A choice of push button or fully automatic water-fill system, allowing the chamber to be topped up after each cycle either with or without operator intervention.
The Uniprogram system is ideal where the same cycle is repeated regularly, the parameters are stored in the memory until altered. Touchclave-R model is versatile and with multi-program control system.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms