Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Freeze drying systems, FreeZone®
Supplier: LABCONCO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7670530 7752030 7670531
535-0033EA 913937 INR InStock
535-0033 535-0037 535-0053
Freeze drying systems, FreeZone®
Freeze Dryers
Designed to handle the lyophilisation needs of research laboratories. Collectors that reach −50 °C are available with capacities from 1 to 18 litres. Cascade systems in 2,5, 4,5, 6 and 12 litre collector capacities that reach −84 °C are ideal for freeze drying samples with low eutectic temperatures. The FreeZone® −105 °C bench top freeze dryer systems are suitable for samples with very low eutectic points including ones containing small amounts of methanol or ethanol.
  • Automatic start-up; pressing one button initiates collector refrigeration and vacuum
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms