Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Rotary vane vacuum pumps
Supplier: LABCONCO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7739403
535-0226EA 0 INR InStock
535-0226
Rotary vane vacuum pumps
Pumps Vacuum Rotary Vane Pumps
This vacuum pump is designed for use with aqueous samples that are compatible with FreeZone Freeze Dry Systems, CentriVap Centrifugal concentrators and Protector® Controlled Atmosphere glove boxes. It comes with a single phase direct drive motor, totally enclosed and fan cooled. When the motor overheats, the thermal overload device switches off the pump. When the pump cools down, the motor automatically restarts. Isolation valve seals the inlet to prevent oil and air contamination of the system in the event of power failure.
  • Two inlet adapters (¹/₂" and ³/₄")
  • Mode selector with two
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms