Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Glove box, stainless steel, filtered, Protector®

Supplier: LABCONCO

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5065537 5065535 5065531 5065533
LBCP5065537EA 0 INR InStock
LBCP5065537 LBCP5065535 LBCP5065531 LBCP5065533
Glove box, stainless steel, filtered, Protector®
Glove Boxes and Glove Bags
These glove boxes with stainless steel liner provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials. Available with two or four glove ports (double version).

  • LCD display, large viewing area (double glove boxes have two viewing areas), space-saving inner and outer transfer chamber doors that pivot upward
  • Two 25-watt fluorescent lights; switches for light and electrical duplex
  • Built-in blower with speed control capable of airflow from 15 CFM to 85 CFM
  • Dry-
More Product Information
Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms