Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Dry evaporation systems, N₂/48, RapidVap®
Supplier: LABCONCO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7910013 7482100 7482200
LBCP7910013EA 0 INR InStock
LBCP7910013 LBCP7482100 LBCP7482200
Dry evaporation systems, N₂/48, RapidVap®
Evaporators Centrifugal Evaporators
This system uses nitrogen blow down, vortex motion and heat to speed evaporation of up to 48 small samples or eight 450 milliliter samples at once.
  • Nitrogen blow down reduces partial pressure directly over the liquid to speed up the evaporation
  • Nitrogen may be introduced to 2, 4, 6 or 8 clusters of 6 sample tubes
  • Microprocessor-controlled 1000-watt dry block heating system supplies a controlled amount of heat up to 100 °C
  • Two-way RS232 link to allow remote control of functions from a user-supplied personal computer
The system may be used to prepareMore Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms