Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Storage cabinets for solvents, Protector®
Supplier: LABCONCO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9902000 9902100 9903000
LBCP9902000EA 280812 INR InStock
LBCP9902000 LBCP9902100 LBCP9903000
Storage cabinets for solvents, Protector®
Furniture Storage Cabinets
These cabinets support fume hoods and safely store and vent solvents and other flammable liquids. The contents inside them are also protected during any fire case.
  • Manual and self closing doors
  • Shelf made of epoxy coated steel
  • Two rear vent connections with flame arrestors and closure plugs
  • Epoxy coated steel construction
Equipped with three-point locking handle, rear vent outlets with flame arresters and closure plugs, dished epoxy-coated steel shelf, and leveling legs. Include one 20,3 cm (8") filler panel to increase cabinet's overall depth from 55,More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms