Respiration BOD: BOD self-checks
Respiration BOD: BOD self-checks
A reduction in oxygen (consumed by micro organisms) causes a definite pressure difference that is measured by a pressure sensor. Practical and easy to perform!