Viton Tubing Specifications
Advantages:
  • Our most chemical-resistant tubing.
  • Resistant to corrosives, solvents, and oils at elevated temperature.
  • Low gas permeability.
Limitations:
  • Limited pumping life.
Application Suitability:
Acids: Excellent
Alkalies: Excellent
Organic solvents: Variable—test before using
Pressure: Good
Vacuum: Good
Viscous fluids: Good
Sterile fluids: Fair
Physical Characteristics and Composition:
  • Thermal set rubber.
  • Viton B (67 percent fluorine).
  • Firm (stiff) material.
  • Opaque, black.
Temperature Range:
-25 to 400°F (-32 to 205°C)
Meets Classifications:
  • None
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2: 76 to 79
H2: -
O2: 13 to 15
N2: 4.3
Cleaning/Sterilization:
  • Sterilize at 480°F (249°C) for 16 hours in a circulating hot air oven.
Viton - Reg TM DuPont Dow Elastomers L.L.C.